Anti-DIP2A/B/C Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-9312R-A750
BOSSBS-9312R-A750EA 893 EUR InStock
BOSSBS-9312R-A750
Anti-DIP2A/B/C Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
DIP2A, also known as DIP2, is a 1571 amino acid nuclear protein. It is one of three human homologs (DIP2A, DIP2B and DIP2C) of the Drosophila dip2 (disconnected-interacting protein 2) protein. In Drosophila, dip2 interacts with disco, a protein required for neuronal connections in the visual systems of larvae and adults. The closest vertebrate homologs to disco are the basonuclin genes. In mice, DIP2 homologs show restricted expression to the brain. This suggests that, similar to the function of Drosophila dip2, vertebrate DIP2 homologs may play a role in the development of the nervous system. Expressed ubiquitously with highest expression in the brain, DIP2A is thought to function in signaling throughout the central nervous system by providing positional clues for axon patterning and pathfinding. Four isoforms of DIP2A exist due to alternative splicing events.
Type: Primary
Antigen: DIP2A/B/C
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/1571
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge