Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DIP2A Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9312R-CY5
BOSSBS-9312R-CY5EA 893 EUR InStock
BOSSBS-9312R-CY5
Anti-DIP2A Rabbit Polyclonal Antibody (Cy5®)
Antikörper
DIP2A, also known as DIP2, is a 1571 amino acid nuclear protein. It is one of three human homologs (DIP2A, DIP2B and DIP2C) of the Drosophila dip2 (disconnected-interacting protein 2) protein. In Drosophila, dip2 interacts with disco, a protein required for neuronal connections in the visual systems of larvae and adults. The closest vertebrate homologs to disco are the basonuclin genes. In mice, DIP2 homologs show restricted expression to the brain. This suggests that, similar to the function of Drosophila dip2, vertebrate DIP2 homologs may play a role in the development of the nervous system. Expressed ubiquitously with highest expression in the brain, DIP2A is thought to function in signaling throughout the central nervous system by providing positional clues for axon patterning and pathfinding. Four isoforms of DIP2A exist due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DIP2A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge