Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LRCH3 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9329R
BOSSBS-9329REA 725 EUR InStock
BOSSBS-9329R
Anti-LRCH3 Rabbit Polyclonal Antibody
Antikörper
Members of the leucine-rich repeat family includes LRCH1, LRCH2, LRCH3 and LRCH4. All family members contain one calponin-homology domain and nine leucine-rich repeats. The best characterized leucine-rich repeat family member is LRCH4, which is suggested to be involved in ligand binding in the brain, with expression observed primarily in the hippocampus. As a cell adhesion molecule and signal receptor, LRCH4 may play an important role in maintenance of hippocampus-dependent memories, with defects in the gene possibly contributing to a loss of long-term memory. The gene encoding LRCH3 maps to human chromosome 3, which spans 200 million base pairs and encodes between 1,100 and 1,500 genes. There are three isoforms of LRCH3 that are produced as a result of alternative splicing events.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: LRCH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge