Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MARCH8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9342R-A680
BOSSBS-9342R-A680EA 893 EUR InStock
BOSSBS-9342R-A680
Anti-MARCH8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. MARCH8 (membrane-associated ring finger (C3HC4) 8), also known as MIR or RNF178 (RING finger protein 178), is a 291 amino acid multi-pass membrane protein that localises to vesicle membranes and contains one RING-CH-type zinc finger. Expressed in a variety of tissues, including immature dendritic cells, MARCH8 functions as an E3 ubiquitin ligase that is thought to regulate immune responses by promoting the ubiquitination and subsequent degradation of target proteins, such as B7-2 and CD71.

Type: Primary
Antigen: MARCH8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/291
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge