Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PSMA6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9355R-A350
BOSSBS-9355R-A350EA 893 EUR InStock
BOSSBS-9355R-A350
Anti-PSMA6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The gene encodes a 246-amino acid polypeptide containing an RNA binding motif, a putative nuclear localization signal, and phosphorylation sites. The alpha subunits comprises the outer rings of the proteasome. Some alpha subunits contain a functional nuclear localization signal; proteasomes are found in both the nuclear and cytoplasmic compartments of the cell. Alpha subunits may constitute a physical barrier that limits access of cytosolic proteins into the inner proteolytic chamber.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PSMA6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge