Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PSMD6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9367R-A488
BOSSBS-9367R-A488EA 893 EUR InStock
BOSSBS-9367R-A488
Anti-PSMD6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
In eukaryotic cells, selective breakdown of cellular proteins is ensured by two distinct pathways, ubiquitination and degradation by the 26S proteasome. At specific stages of development, embryo- and tissue-specific components of the 26S proteasome are formed by developmentally regulated alternative splicing, including Rpn10a through Rpn10e (also designated pUb-R2 through pUb-R5). The pUb-R2 subunit, originally identified as S5a, is ubiquitously expressed and may perform proteolysis constitutively in a wide variety of cells. p44S10 is a highly conserved proteasome regulatory subunit that is expressed in heart, liver, skeletal muscle and pancreas. In addition to normal tissue expression, p44S10 is also expressed in several melanoma cell lines, such as MCF-7, 451Lu and WM164. Since forced expression of p44S10 in radial growth phase melanoma cells results in an increase in cellular proliferation, p44S10 may represent a potential link between regulation of proteasome activity and tumor cell proliferation in vivo.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PSMD6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge