Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TRIM54 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9376R
BOSSBS-9376REA 725 EUR InStock
BOSSBS-9376R
Anti-TRIM54 Rabbit Polyclonal Antibody
Antikörper
MuRF1, is a nuclear protein that interacts with SMT3b and the large myofibrillar protein Titin. In muscle cells, MuRF2 (RFN29) regulates gene expression and protein turnover. It localizes to the cytoplasm, but under atrophic conditions it is detected in the nucleus. MuRF2 can form oligomers with various other proteins, including Titin and Myosin. MuRF3, also designated tripartite motif-containing 54 (TRIM54) or ring finger protein 30 (RNF30), interacts with tubulin and stabilizes microtubules duing myotube formation. It is a cytoplasmic protein the localizes to the Z-lines in skeletal muscles, while MuRF2 localizes to the sarcomeric M-band in cardiomyocytes. MuRF3 shares 77% and 65% sequence identity with MuRF1 and MuRF2, respectively. MuRF1-3 share a conserved N-terminal RING domain and zinc-binding B-box motif, and two coiled-coil dimerization motif boxes, in their central regions.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: TRIM54
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge