Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MKRN2 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9379R
BOSSBS-9379REA 725 EUR InStock
BOSSBS-9379R
Anti-MKRN2 Rabbit Polyclonal Antibody
Antikörper
The Makorins are a family of putative ribonucleoproteins containing two to four C3H zinc fingers that may confer RNA-binding. In addition, they contain a C3HC4 RING zinc finger that allows them to function as E3 ubiquitin ligases. Makorin-2, also known as RNF62, HSPC070 or MKRN2, is a widely expressed, evolutionarily conserved protein with four C3H-type zinc fingers (three at the N-terminus and one at the C-terminus), one RING-type zinc finger and a cysteine and histidine motif similar to that found in Makorin-1. In Xenopus, Makorin-2 functions, via PI 3-kinase/Akt signaling, as a negative regulator of neurogenesis. In humans, Makorin-2 is overexpressed in various cancer cell lines, suggesting a possible role of Makorin-2 in tumor progression. In addition, Makorin-2 is co-expressed with Raf-1 in the same tissues and cell lines.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: MKRN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge