Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CNOT2 Ser101 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9384R-A750
BOSSBS-9384R-A750EA 1350 EUR InStock
BOSSBS-9384R-A750
Anti-CNOT2 Ser101 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
CNOT2 (CCR4-NOT transcription complex subunit 2) is a ubiquitous protein encoded by the human gene CNOT2. CNOT2 belongs to the CNOT2/3/5 family and is part of the CCR4-NOT complex. The CCR4-NOT complex is an evolutionarily conserved, multi-component complex known to be involved in transcription as well as mRNA degradation. Various subunits (e.g. CNOT1, CNOT2) are involved in influencing nuclear hormone receptor activities. The CCR4-NOT complex is also involved in the regulation of Histone H3 lysine 4 methylation through a ubiquitin-dependent pathway that likely involves the proteasome. Increased expression of the CNOT2 subunit acts to strongly repress transcription by RNA polymerase II. This repressive effect is mediated by a conserved NOT-Box, which is located at the C-terminus of CNOT2 proteins. Repression by the NOT-Box is sensitive to treatment with the histone deacetylase (HDAC) inhibitor trichostatin A.

Type: Primary
Antigen: CNOT2 Ser101
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge