Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CAF1p55 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9393R-A750
BOSSBS-9393R-A750EA 893 EUR InStock
BOSSBS-9393R-A750
Anti-CAF1p55 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
P55 is an extensively palmitoylated erythrocyte membrane protein, and a member of the MAGUK family. P55 also resists salt extraction, resulting in a high affinity for the plasma membrane. P55 contains a PDZ/DHR domain, a conserved SH-3 domain that appears to suppress tyrosine kinase activity of various oncoproteins, a 39-amino acid motif that binds to cytoskeletal protein 4.1R, and a guanylate kinase-like domain. Interaction with glycophorin C (GPC) and 4.1R suggests that p55 may play a role in the dynamic regulation in the erythrocyte membrane. In addition, p55 gene expression in vivo may be associated with a CpG island. P55 is constitutively expressed in K562 erythroleukemia cells during erythropoiesis and undergoes a 2-fold amplification after induction.

Type: Primary
Antigen: CAF1p55
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 221-320/1507
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge