Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DCAF12 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9394R-CY5.5
BOSSBS-9394R-CY5.5EA 900 EUR InStock
BOSSBS-9394R-CY5.5
Anti-DCAF12 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
WD-repeats are motifs that are found in a variety of proteins and are characterized by a conserved core of 40-60 amino acids that commonly form a tertiary propeller structure. While proteins that contain WD-repeats participate in a wide range of cellular functions, they are generally involved in regulatory mechanisms concerning chromatin assembly, cell cycle control, signal transduction, RNA processing, apoptosis and vesicular trafficking. WDR40A (WD-repeat-containing protein 40A), also known as DDB1- CUL4-associated factor 12, is a 453 amino acid cytoplasmic protein that contains four WD-repeats. WDR40A is highly expressed in some cancer cell lines, lung cancer tissues and normal testis. A probable substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex, WRD40A is suggested to interact with DDB1. WDR40A is encoded by a gene mapping to human chromosome 9p13.3. Human chromosome 9 houses over 900 genes and comprises nearly 4% of the human genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DCAF12
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge