Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ODF3 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9406R
BOSSBS-9406REA 725 EUR InStock
BOSSBS-9406R
Anti-ODF3 Rabbit Polyclonal Antibody
Antikörper
Outer dense fibers are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail. May help to maintain the passive elastic structures and elastic recoil of the sperm tail.Constituting the main cytoskeletal structure of spermatid flagella, outer dense fibers (ODFs) add elastic recoil, stiffness and protection against shear forces during sperm movement. Human ODFs consist of approximately 10 major and at least 15 minor proteins. The major proteins of the ODF include Odf1, Odf2, and Odf3, which compose a family of proteins that are preferentially expressed during mammalian spermiogenesis. Odf3 (outer dense fiber protein 3), also known as sperm tail protein SHIPPO 1 and TISP50 (transcript induced in spermiogenesis protein 50), is a 254 amino acid protein that is expressed during the latter part of spermatogenesis in flagella of elongated spermatids and mature sperm. Odf proteins are directed to their exact subcellular location by Spags, which are characterized as chaperone-like Odf-binding molecules. There are two isoforms of Odf3 that are produced as a result of alternative splicing events.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ODF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge