Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-OIF Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9407R-A750
BOSSBS-9407R-A750EA 893 EUR InStock
BOSSBS-9407R-A750
Anti-OIF Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The small leucine-rich proteoglycan (SLRP) family of proteins contains various proteins such as Decorin, Biglycan, Fibromodulin, Keratocan, Lumican, Osteoadherin and Osteoglycin. These proteins all have similar functions as they all mediate extracellular matrix organization and act as binding partners of TGF beta. Osteoglycin, which also may be designated osteoinductive factor (OIF), is a secreted protein detected in bone tissues. Osteoglycin induces the formation of bone in conjunction with either TGF-beta-1 or TGF-beta-2. The precursor form of the OGN gene product, designated Mimecan, is subject to in situ proteolytic cleavage to yield the mature Osteoglycin.

Type: Primary
Antigen: OIF
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/298
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge