Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF27 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9432R-A680
BOSSBS-9432R-A680EA 893 EUR InStock
BOSSBS-9432R-A680
Anti-RNF27 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The tripartite motif (TRIM) family of proteins are characterised by a conserved TRIM domain that includes a coiled-coil region, a B-box type zinc finger, one RING finger and three zinc-binding domains. TRIM8 (tripartite motif containing 8), also known as GERP (glioblastoma-expressed RING finger protein) or RNF27 (RING finger protein 27), is a 551 amino acid protein that is thought to function as an E3 ubiquitin-protein ligase that promotes SOCS-1 proteasomal degradation. As a widely expressed homodimer, TRIM8 localizes to nuclear bodies and contains two B box-type zinc fingers and one RING-type zinc finger. TRIM8 is expressed in lung, heart, brain and skeletal muscle, with low levels detected in intestine, placenta, leukocytes and liver. The gene encoding TRIM8 maps to human chromosome 10q24.32.

Type: Primary
Antigen: RNF27
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 61-160/551
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge