Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PAPOL A+B+G Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9435R-CY7
BOSSBS-9435R-CY7EA 900 EUR InStock
BOSSBS-9435R-CY7
Anti-PAPOL A+B+G Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Polyadenylation of the 3-prime ends of eukaryotic mRNAs is a key event that takes place in the nucleus during maturation of mRNA. The reaction includes endoribonucleolytic cleavage of the pre-RNA at the poly(A) site that leads to synthesis of the poly(A) tail at the 3-prime end of the upstream cleavage product. The poly(A) polymerase (PAP) is required The adenosine addition reaction depends on poly(A) polymerase (PAP) activity. The testis express PAP-beta (TPAP) in the cytoplasm of spermatogenic cells. The adenosine addition function of PAP-beta plays a critical role in male germ cell production. PAP-beta-deficient transgenic mice display impaired expression of haploid-specific genes that are necessary for spermatogenesis. The intronless gene encoding human PAP-beta maps to chromosome 7p22.3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAPOL A+B+G
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge