Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MTA2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9440R-A555
BOSSBS-9440R-A555EA 893 EUR InStock
BOSSBS-9440R-A555
Anti-MTA2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
MTA1 is a component of the NURD (nucleosome remodeling and histone deacetylation) complex, which is associated with ATP-dependent chromatin-remodeling and histone deacetylase activity. MTA1 functions in conjunction with other components of NURD to mediate transcriptional repression as it facilitates the association of repressor molecules with the chromatin. Structurally, MTA1 contains a single SH3-binding motif and a zinc finger domain, along with a region similar to the co-repressor protein N-Cor. MTA1 is normally expressed at low levels in various tissues and is more highly expressed in testis. Overexpression of MTA1 correlates with tumor invasion and metastasis in various carcinomas including colorectal, gastrointestinal and breast carcinomas. Elevation of MTA1 levels in these tumors appears to enhance the metastases to lymph nodes, increase mammary cell motility and potentiate growth, and therefore may be an indicator for assessing the potential malignancies of various tumors. A similar protein, MTA2, also designated MTA1-L1 (MTA1-like protein 1), shares more than 55% sequence homology with MTA1 and is ubiquitously expressed.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MTA2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge