Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TNNI3K Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9458R-FITC
BOSSBS-9458R-FITCEA 900 EUR InStock
BOSSBS-9458R-FITC
Anti-TNNI3K Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
TNNI3K, also known as CARK, is a 936 amino acid serine/threonine-protein kinase that is highly expressed in heart. Overexpression of TNNI3K leads to improved cardiac function by enhancing beating frequency and increasing contractile force and epinephrine response. TNNI3K suppresses phosphorylation of cardiac troponin I and p38/JNK-mediated apoptosis, therefore protecting the myocardium from ischemic injury. Administration of TNNI3K to mice with myocardial infarction improves cardiac performance and attentuates ventricular remodeling, suggesting that TNNI3K could be a promising target in the treatment of cardiac diseases. There are four isoforms of TNNI3K that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNNI3 interacting kinase
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge