Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HEY2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9461R-CY5.5
BOSSBS-9461R-CY5.5EA 900 EUR InStock
BOSSBS-9461R-CY5.5
Anti-HEY2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The LIN-12/Notch family of transmembrane receptors plays a central role in development by regulating cell fate and establishing boundaries of gene expression. Notch signaling activates the Hairy/Enhancer of split (HES) genes, which encode basic helix-loop-helix (bHLH) transcriptional repressors that are critical for directing embryonic patterning and development. The Hairy-related transcription factors (HRTs) comprise a subclass of bHLH proteins that exhibit structural similarity with the HES proteins and include HRT1, HRT2 and HRT3. The HRT family (also designated Hesr, Hey, CHF and Gridlock) contain a bHLH domain, an Orange domain and a novel YRPW domain, which is absent in HRT3. The Hairy-related genes map to human chromosomes 8q21, 6q21 and 1p34.3 for HRT1, HRT2 and HRT3, respectively, and are downstream targets for Notch signaling. HRT1 is expressed in the somitic mesoderm, central nervous system, kidney, heart, nasal epithelium and limb buds in murine embryos as well as in adult tissues. It has altered expression in many breast, lung and kidney tumors. Like HRT1, HRT2 and HRT3 are also expressed in developing somites, heart and nervous system.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HEY2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge