Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SMARCD3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9479R-CY5.5
BOSSBS-9479R-CY5.5EA 900 EUR InStock
BOSSBS-9479R-CY5.5
Anti-SMARCD3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
SMARCD3, is a member of the SMARCD family and contains one SWIB domain. Two isoforms, isoform 1 and isoform 2 exist due to alternative splicing events. Both isoforms are expressed in placenta, salivary gland, kidney, brain, trachea, uterus, prostate, testis, thyroid, spleen and heart, while isoform 1 is also expressed in adipose tissue and skeletal muscle. Localizing to the nucleus, SMARCD3 is a component of the ATP-dependent chromatin remodeling complex SNF/SWI and is believed to play a role in nucleosome remodeling. SMARCD3 also plays an important role in the regulation of muscle development. In mice, the silencing of the gene en-coding SMARCD3 leads to defects in heart morphogenesis. In addition, both isoforms of SMARCD3 directly interact with and function as coactivators for several transcription factors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SMARCD3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge