Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-F9 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9500R-CY3
BOSSBS-9500R-CY3EA 897 EUR InStock
BOSSBS-9500R-CY3
Anti-F9 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Hemostasis following tissue injury involves the deployment of essential plasma procoagulants (prothrombin, and factors X, IX, V, and VIII), which are involved in a blood coagulation cascade that leads to the formation of insoluble fibrin clots and the promotion of platelet aggregation (1-3). Coagulation factor IX (plasma thromboplastic component, F9, F.IX, HEMB) is a vitamin K-dependent, single chain serine protease that is synthesized in the liver and circulates as an inactive precursor (3,4). Factor XIa mediated proteolytic cleavage of factor IX generates factor IXa, an active serine protease composed of a 145 amino acid light chain and a 236 amino acid catalytic heavy chain, linked through disulfide bonds (5). Genetic alterations at the Factor IX locus such as point mutations, insertions and deletions, can lead to hemophilia B, also known as Christmas disease (6).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Factor IX
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge