Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-F10 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9501R-CY5
BOSSBS-9501R-CY5EA 900 EUR InStock
BOSSBS-9501R-CY5
Anti-F10 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Hemostasis following tissue injury involves the deployment of essential plasma procoagulants (Prothrombin and Factors X, IX, V and VIII), which are involved in a blood coagulation cascade that leads to the formation of insoluble Fibrin clots and the promotion of platelet aggregation. Coagulation Factor X (Stuart Prower factor, FX, F10) is a vitamin K-dependent, single chain serine protease that is synthesized in the liver and circulates as an inactive precursor. The mature form of Factor X (Factor X A) is generated by Factor IX A- or Factor VII A-mediated cleavage at the tripeptide sequence, Arg-Lys-Arg, to yield a disulfide linked dimer. Together with the cofactor Factor V A and Ca2+ on the surface of platelets or endothelial cells, Factor X A coordinates as part of the prothrombinase complex, which mediates proteolysis of Prothrombin into active Thrombin. Mutations at the Factor X locus resulting in Factor X deficiencies can contribute to hemorrhagic diathesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: F10
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge