Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LRRC3B Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9509R-A350
BOSSBS-9509R-A350EA 893 EUR InStock
BOSSBS-9509R-A350
Anti-LRRC3B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Leucine-rich repeats (LRRs) are 20-29 amino acid motifs that mediate protein-protein interactions. The primary function of these motifs is to provide a versatile structural framework for the formation of these protein-protein interactions. LRRs are present in a variety of proteins with diverse structure and function, including innate immunity and nervous system development. Several human diseases are associated with mutation in the genes encoding LRR-containing proteins. The leucine-rich repeat-containing protein 3B (LRRC3B), also designated LRP15, is a 259 amino acid protein that contains 3 LRR repeats. The gene encoding LRRC3B is a tumor suppressor gene that is regulated by DNA methylation. Decreased expression of LRRC3B has been shown in colorectal cancer and gastric cancer, making LRRC3B a candidate marker for those cancers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRRC3B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge