Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BPGM Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9519R-A350
BOSSBS-9519R-A350EA 893 EUR InStock
BOSSBS-9519R-A350
Anti-BPGM Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
BPGM (2,3-bisphosphoglycerate mutase) is a 259 amino acid protein that belongs to the phosphoglycerate mutase family and exists as a homodimer that plays a crucial role in the regulation of hemoglobin oxygen. Specifically, BPGM catalyzes the conversion of 3-D-glyceroyl phosphate to 2,3-bisD-glycerate (2,3-BPG), a reaction that is essential for controlling the concentration of 2,3-BPG within the cell. The gene encoding BPGM maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. Involvement in disease:Defects in BPGM are the cause of bisphosphoglycerate mutase deficiency (BPGMD) . A disease characterized by hemolytic anemia, splenomegaly, cholelithiasis and cholecystitis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BPGM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge