Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MPP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9522R-HRP
BOSSBS-9522R-HRPEA 900 EUR InStock
BOSSBS-9522R-HRP
Anti-MPP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The Kinesins constitute a large family of microtubule-dependent motor proteins which are responsible for the distribution of numerous organelles, vesicles and macromolecular complexes throughout the cell. Individual Kinesin members play crucial roles in cell division, intracellular transport and membrane trafficking events, including endocytosis and transcytosis. MPP1 (M-phase phosphoprotein 1), also known as KIF20B (kinesin family member 20B), MPHOSPH1 or KRMP1, is a 1,820 amino acid protein that localizes to both the nucleus and the cytoplasm and contains one kinesin-motor domain. Expressed in kidney, brain, testis and ovary, MPP1 functions as a plus-end directed motor enzyme that interacts with Pin1 and is required for the completion of cytokinesis. MPP1, which exists as multiple alternatively spliced isoforms termed 1-5, is subject to post-translational phosphorylation, probably by ATM or ATR.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: MPP1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge