Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ORAI2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9541R-CY5.5
BOSSBS-9541R-CY5.5EA 900 EUR InStock
BOSSBS-9541R-CY5.5
Anti-ORAI2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Orai2 is a 254 amino acid multi-pass membrane protein that belongs to the orai family of proteins. Localizing to the plasma membrane, Orai2 plays an important role in store-operated calcium (SOC) entry, a process involving Ca2+ influx and replenishment of Ca2+ stores formerly emptied through the action of inositol 1,4,5-trisphosphate production and other Ca2+ mobilizing agents. CRAC channels are responsible for medi-ating calcium influx in T-cells and play an important role in the immune response. Orai2 specifically increases the Ca2+-selective current through coaction with the Ca2+ sensor Stim1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ORAI2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge