Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BEND2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9551R-CY3
BOSSBS-9551R-CY3EA 900 EUR InStock
BOSSBS-9551R-CY3
Anti-BEND2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
BEND2 is a 799 amino acid protein that contains two BEN domains. BEND2 exists as two alternatively spliced isoforms and is considered a complete proteome. BEN domain mediates protein–DNA and protein–protein interactions during chromatin organization and transcription. BEN domain may play a role in organization of viral DNA during replication or transcription. The BEND2 gene maps to human chromosome Xp22.13. Chromosome X consists of about 153 million base pairs and nearly 1,000 genes. The X and Y chromosomes are the human sex chromosomes. The combination of an X and Y chromosome lead to normal male development while two copies of X lead to normal female development. Color blindness, hemophilia, and Duchenne muscular dystrophy are well known X chromosome-linked conditions which affect males more frequently as males carry a single X chromosome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BEND2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge