Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GLTPD2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9557R-A680
BOSSBS-9557R-A680EA 893 EUR InStock
BOSSBS-9557R-A680
Anti-GLTPD2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The GLTP (glycolipid transfer protein) superfamily is defined by a unique lipid transfer/binding fold (GLTP fold) that accelerate glycolipid intermembrane transfer. GLTPD2 is a 291 amino acid protein that belongs to the GLTP family. The gene encoding GLTPD2 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1200 genes. Two key tumour suppressor genes are associated with chromosome 17, p53 and BRCA1. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome. Neurofibromatosis, a condition characterised by neural and epidermal lesions, and dysregulated Schwann cell growth are both linked to mutations on chromosome 17. Alexander disease, Birt-Hogg-Dube syndrome and Canavan disease are also associated with chromosome 17.

Type: Primary
Antigen: GLTPD2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 171-270/291
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge