Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SHARPIN Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9581R-CY5.5
BOSSBS-9581R-CY5.5EA 900 EUR InStock
BOSSBS-9581R-CY5.5
Anti-SHARPIN Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
SHARPIN is a 387 amino acid protein that localizes to the cytoplasm and contains one RanBP2-type zinc finger. Expressed at high levels in placenta and skeletal muscle and present at lower levels in colon, brain, heart, liver, kidney, lung, thymus and small intestine, SHARPIN interacts with Shank 1 and is thought to play a role in the control of inflammatory responses and in the overall development of the immune system. SHARPIN exists as three alternatively spliced isoforms and shares 73% sequence identity with its mouse counterpart, suggesting a conserved role between species. The gene encoding SHARPIN maps to human chromosome 8, which consists of nearly 146 million base pairs, houses more than 800 genes and is associated with a variety of diseases and malignancies.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SHARPIN
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge