Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CRYGS Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9585R-A555
BOSSBS-9585R-A555EA 893 EUR InStock
BOSSBS-9585R-A555
Anti-CRYGS Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Crystallins are water soluble structural proteins found in the vertebrate eye. Mammalian crystallins are classified in three forms, designated α, β and γ. Crystallins, as the principal components of the lens, function to increase the refractive index of the eye during accommodation by forming high-molecular weight aggregates which maintain transparency. γS-crystallin (Gamma-crystallin S), also known as Beta-crystallin S, is a 178 amino acid protein that exists as a monomer which does not aggregate. γS-crystallin contains a two-domain beta structure and belongs to the beta/gamma-crystallin gene family mapping to human chromosome 3. γS-crystallin has been linked to congenital cataract development, a disorder signified by increasing levels of lens opacity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: gamma crystallin S
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge