Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SPAG17 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9596R
BOSSBS-9596REA 725 EUR InStock
BOSSBS-9596R
Anti-SPAG17 Rabbit Polyclonal Antibody
Antikörper
SPAG17 (sperm associated antigen 17), also known as PF6, is a 2,223 amino acid cytoplamic protein that colocalizes with SPAG6 to microtubles. Highly expressed in testis and in organs that contain cilia-bearing cells including brain, oviduct, lung, and uterus, SPAG17 may be important for the structural integrity of the central apparatus of the sperm axoneme. SPAG17 contains two LRR (leucine-rich) repeats and may also participate in flagellar motility and male fertility.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SPAG17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge