Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TEF3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9603R-A488
BOSSBS-9603R-A488EA 893 EUR InStock
BOSSBS-9603R-A488
Anti-TEF3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
TEF-3, is a 427 amino acid member of the transcriptional enhancer factor (TEF) family of proteins that are characterized by the presence of a TEA DNA-binding domain. Localized to the nucleus and expressed primarily in skeletal muscle, TEF-3 functions as a transcriptional regulator by binding specifically and non-cooperatively to the M-CAT motif found in the promotors of muscle-specific genes, thereby directing their subsequent expression. TEF-3 contains one TEA DNA-binding domain and is expressed as multiple isoforms due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TEF3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge