Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C14ORF106 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9615R-FITC
BOSSBS-9615R-FITCEA 900 EUR InStock
BOSSBS-9615R-FITC
Anti-C14ORF106 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
C14orf106 is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis. It interacts with SP1. There are two isoforms.Chromosome 14 contains about 700 genes and 106 million base pairs and makes up about 3.5% of human cellular DNA. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease. The SERPINA1 gene is located on chromosome 14 and when defective leads to the genetic disorder ?-antitrypsin deficiency. This disorder is characterized by severe lung complications and liver dysfunction. Notably, the immunoglobulin heavy chain locus is found on chromosome 14 and has been identified as a fusion with the chromosome 19 encoded protein BCL3 in the (14;19) translocations found in a variety of B cell malignancies. The C14orf106 gene product has been provisionally designated C14orf106 pending further characterization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: C14ORF106
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge