Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-USB1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9631R-CY5
BOSSBS-9631R-CY5EA 900 EUR InStock
BOSSBS-9631R-CY5
Anti-USB1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Involvement in disease;Defects in C16orf57 are the cause of poikiloderma with neutropenia (PN). PN is a genodermatosis characterized by poikiloderma, pachyonychia and chronic neutropenia. The disorder starts as a papular erythematous rash on the limbs during the first year of life. It gradually spreads centripetally and, as the papular rash resolves, hypo- and hyperpigmentation result, with development of telangiectasias. Another skin manifestation is pachyonychia, but alopecia and leukoplakia are distinctively absent. One of the most important extracutaneous symptoms is an increased susceptibility to infections, mainly affecting the respiratory system, primarily due to a chronic neutropenia and to neutrophil functional defects. Bone marrow abnormalities account for neutropenia and may evolve into myelodysplasia associated with the risk of leukemic transformation. Poikiloderma with neutropenia shows phenotypic overlap with Rothmund-Thomson syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: USB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge