Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PDZD9 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9632R-A750
BOSSBS-9632R-A750EA 893 EUR InStock
BOSSBS-9632R-A750
Anti-PDZD9 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
PDZD9 (PDZ domain containing 9) is a 264 amino acid protein that contains one PDZ (DHR) domain and participates in protein binding. Conserved in chimpanzee, dog, cow, mouse and rat, PDZD9 exists as two alternatively spliced isoforms and is encoded by a gene that maps to human chromosome 16p12.1. Chromosome 16 encodes over 900 genes, approximately 90 million base pairs, makes up nearly 3% of human cellular DNA and is associated with a variety of genetic disorders. Giant axonal neuropathy, a nervous system disorder characterised by increasing malfunction with growth, and the rare disorder Rubinstein-Taybi syndrome, characterised by mental retardation and predisposition to tumour growth and white blood cell neoplasias, are associated with chromosome 16. Crohn's disease, a gastrointestinal inflammatory condition, and systemic lupus erythematosis are also associated with chromosome 16.

Type: Primary
Antigen: PDZD9
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/264
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge