Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IDH1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9634R-A350
BOSSBS-9634R-A350EA 893 EUR InStock
BOSSBS-9634R-A350
Anti-IDH1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The Isocitrate dehydrogenase cytoplasmic enzyme is a homodimer of 416 residues that belongs to the isocitrate and isopropylmalate dehydrogenases family. IDHC catalyzes the third step of the citric acid cycle, which involves the oxidative decarboxylation of isocitrate, forming ?ketoglutarate and CO2 in a two step reaction. The first step involves the oxidation of isocitrate to the intermediate oxalosuccinate, while the second step involves the production of ?ketoglutarate. During this process, either NADH or NADPH is produced along with CO2. Ca2+ can bind to IDHC as a complex with isocitrate, acting as a competitive inhibitor of Mg2+. The IDHC enzyme is inactivated by phosphorylation at Ser-113 and contains a clasp-like domain wherein both polypeptide chains in the dimer interlock. IDHC is expressed in a wide range of species and also in organisms that lack a complete citric acid cycle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IDH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge