Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ASH1L Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9671R-A555
BOSSBS-9671R-A555EA 893 EUR InStock
BOSSBS-9671R-A555
Anti-ASH1L Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
ASH1L is a 2,969 amino acid protein encoded by the human gene ASH1L. ASH1L belongs to the histone-lysine methyltransferase family (SET2 subfamily) and contains three AT hook DNA-binding domains, one AWS domain, one BAH domain, one bromodomain, one PHD-type zinc finger, one post-SET domain and one SET domain. It is a widely expressed nuclear protein with highest expression found in brain, heart and kidney. ASH1L is a histone methyltransferase and is believed to methylate 'Lys-4' of Histone H3, which is a specific tag for epigenetic transcriptional activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ASH1L
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge