Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FAM29A Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9723R-CY5.5
BOSSBS-9723R-CY5.5EA 900 EUR InStock
BOSSBS-9723R-CY5.5
Anti-FAM29A Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The human augmin complex (HAUS) is an evolutionarily conserved 8-subunit protein complex that was initially discovered in Drosophila. The HAUS complex is essential for microtubule generation, centrosome integrity, mitotic spindle assembly and completion of cytokinesis. HAUS6 (HAUS augmin-like complex, subunit 6), also known as FAM29A or Dgt6, is a 955 amino acid component of the augmin complex. Required for mitotic progression, HAUS6 localizes to cytoplasm, cytoskeleton, mitotic spindle microtubules and interphase centrosomes, and undergoes post-translational phosphorylation following mitosis on multiple serine and threonine residues. HAUS6 exists as two alternatively spliced isoforms that are encoded by a gene located on human chromosome 9.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM29A
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge