Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TSEN2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9735R-A488
BOSSBS-9735R-A488EA 893 EUR InStock
BOSSBS-9735R-A488
Anti-TSEN2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The tRNA-splicing endonuclease complex is responsible for identifying and cleaving pre-tRNA at both 5’ and 3’ splice sites, thereby releasing introns and free tRNA molecules with 2',3' cyclic phosphates and 5'-OH termini. In addition to its role in pre-tRNA splicing, the heterotetrameric endonuclease complex participates in mRNA processing and, via its association with pre-mRNA processing factors, is thought to link pre-tRNA and pre-mRNA splicing events. TSEN2 (tRNA-splicing endonuclease subunit Sen2), also known as tRNA-intron endonuclease Sen2, is a 465 amino acid nuclear protein that constitutes one of the two catalytic subunits of the tRNA-splicing endonuclease complex. There are three isoforms of TSEN2 that are produced as a result of alternative splicing events. Isoform 1 seems to carry the active site for 5’-splice site cleavage. Defects in the gene encoding TSEN2 are the cause of pontocerebellar hypoplasia type 2B, which is characterized by progressive microencephaly with epilepsy, extrapyramidal dyskinesia and chorea without spinal cord findings.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSEN2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge