Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EDRF1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9737R-CY3
BOSSBS-9737R-CY3EA 900 EUR InStock
BOSSBS-9737R-CY3
Anti-EDRF1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The tetratricopeptide repeat (TPR) motif is a degenerate, 34 amino acid sequence found in many proteins and acts to mediate protein-protein interactions in various pathways. At the sequence level, there can be up to 16 tandem TPR repeats, each of which has a helix-turn-helix shape that stacks on other TPR repeats to achieve ligand binding specificity. EDRF1 (erythroid differentiation-related factor 1), also known as C10orf137 (chromosome 10 open reading frame 137), is a 1,238 amino acid protein containing two TPR repeats. Localizing to nucleus, EDRF1 is involved in transcriptional activation of globin genes by regulating DNA-binding activity of GATA-1 transcription factor. EDRF1 may also play an important role in organ development and histological differentiation. EDRF1 exists as four alternatively spliced isoforms and is encoded by a gene mapping to human chromosome 10q26.13.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EDRF1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge