Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ANGEL1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9741R-CY5
BOSSBS-9741R-CY5EA 900 EUR InStock
BOSSBS-9741R-CY5
Anti-ANGEL1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
ANGEL1 , also known as KIAA0759, is a 670 amino acid protein belonging to the CKR-4 protein family. ANGEL1 is encoded by a gene located on human chromosome 14, which contains about 700 genes and 106 million base pairs, making up about 3.5% of human cellular DNA. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of three key genes associated with the development of Alzheimer's disease. The SERPINA1 gene is located on chromosome 14 and, when defective, leads to the genetic disorder ?-antitrypsin deficiency. This disorder is characterized by severe lung complications and liver dysfunction. Notably, the immunoglobulin heavy chain locus on chromosome 14 and its fusion via translocation with the chromosome 19 encoded protein BCL3 may be related to B-cell malignancies.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANGEL1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge