Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C10orf4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9767R-A680
BOSSBS-9767R-A680EA 893 EUR InStock
BOSSBS-9767R-A680
Anti-C10orf4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
C10orf4, also known as FRA10A or FRA10AC1, is a 315 amino acid nuclear protein that is expressed strongly in kidney, liver, heart, brain and skeletal muscle. Defects in the gene encoding C10orf4 are characterised by an expansion of a polymorphic CGG repeat within the 5'-UTR of the gene; an event that is thought to cause folate-sensitive fragile site FRA10A expression. Expression of this mutated FRA10A protein may be associated with tumourigenesis, neurological disorders and mental retardation. C10orf4 is believed to be conserved between species. Five isoforms of C10orf4, all of which differ in their C-termini, are expressed due to alternative splicing events.

Type: Primary
Antigen: C10orf4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/315
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge