Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C10orf63 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9773R-A680
BOSSBS-9773R-A680EA 893 EUR InStock
BOSSBS-9773R-A680
Anti-C10orf63 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Enkurin is a 256 amino acid adapter protein that brings signal transduction proteins and transient receptor potential canonical (TRPC) cation channels into contact. Localizing to the acrosomal crescent and flagellar principal piece of sperm, Enkurin contains one IQ domain which it uses to bind CaM (calmodulin). Enkurin is highly expressed in testis and vomeronasal organ, but is also found at lower levels in brain, ovary, heart and lung. The gene encoding Enkurin maps to human chromosome 10, which contains over 800 genes and 135 million nucleotides. PTEN is an important tumour suppressor gene located on chromosome 10 and, when defective, causes a genetic predisposition to cancer development known as Cowden syndrome. Other chromosome 10 associated disorders include Cockayne syndrome, tetrahydrobiopterin deficiency and trisomy 10.

Type: Primary
Antigen: C10orf63
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 181-256/256
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge