Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C1orf25 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9784R-A680
BOSSBS-9784R-A680EA 893 EUR InStock
BOSSBS-9784R-A680
Anti-C1orf25 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
C1orf25, also known as TRM1L (TRMT1-like protein), MST070 or MSTP070, is a 733 amino acid protein that belongs to the TRM1 family of proteins. Expressed in a wide variety of tissues, C1orf25 contains one C2H2-type zinc finger and shares sequence similarity with TRMT1, a tRNA methyltransferase that dimethylates guanine at position 26 of tRNAs. As is suggested by observations of C1orf25-deficient mice, C1orf25 is believed to play a role in exploratory behavior and motor coordination. The gene encoding C1orf25 localizes to chromosome 1 and, due to alternative splicing events, C1orf25 exists as at least two isoforms.

Type: Primary
Antigen: C1orf25
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 631-733/733
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge