Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AE2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9847R-A750
BOSSBS-9847R-A750EA 1350 EUR InStock
BOSSBS-9847R-A750
Anti-AE2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Primary canalicular bile undergoes a process of fluidization and alkalinization along the biliary tract that is influenced by several factors, including hormones, innervation/neuropeptides and biliary constituents. The excretion of bicarbonate at both the canaliculi and the bile ducts is an important contributor to the generation of bile-salt independent flow. Bicarbonate is secreted from hepatocytes and cholangiocytes through parallel mechanisms, which involve chloride efflux through activation of chloride channels and further bicarbonate secretion via AE2 (also designated SLC4A2)-mediated chloride/bicarbonate exchange. The AE2 protein regulates pH, chloride concentration, cell volume and transepithelial ion transport in many tissues. Gene silencing of AE2 causes a marked inhibition of unstimulated and secretin-stimulated chloride/bicarbonate exchange, which maintains the bile acid pool that is crucial for secretin to induce bicarbonate-rich choleresis.

Type: Primary
Antigen: AE2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 851-950/1241
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge