Anti-Cyclophilin 40 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-9878R-A680
BOSSBS-9878R-A680EA 893 EUR InStock
BOSSBS-9878R-A680
Anti-Cyclophilin 40 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Cyclophilins are conserved, ubiquitous and abundant cytosolic peptidyl-prolyl cis-trans isomerases that accelerate the isomerization of XaaPro peptide bonds and the refolding of proteins. Human cyclophilin A (CyPA), an intracellular protein of 165 amino acids, is the target of cyclosporin A (CsA) and is encoded by a single unique gene conserved from yeast to humans. Cyclophilin B (CyPB) is secreted in biological fluids such as blood or milk and binds to a specific receptor present on the human lymphoblastic cell line Jurkat and on human peripheral blood lymphocytes. Cyclophilin D (CyP40) is a widely expressed cytoplasmic protein that catalyses the cis-trans isomerization of proline imidic peptide bonds in oligopeptides. It is a widely expressed cytoplasmic protein.
Type: Primary
Antigen: Cyclophilin 40
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 281-370/370
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge