Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HEBP1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9887R-A750
BOSSBS-9887R-A750EA 893 EUR InStock
BOSSBS-9887R-A750
Anti-HEBP1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
p22HBP, also known as HEBP1 (heme binding protein 1), HBP or HEBP, is a 189 amino acid intracellular tetrapyrrole-binding protein that assists in prevention of cellular toxicity by removing free porphyrinogens from the cell. Existing as a monomer, p22HBP localizes to cytoplasm and contains a 21 amino acid chemoattractant within its N-terminus that functions as a natural ligand for FPR3. p22HBP is a member of the HEBP family and binds N-methylprotoporphyrin and metalloporphyrins with similar affinity to porphyrinogens. The gene encoding p22HBP maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.

Type: Primary
Antigen: HEBP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/189
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge