Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PGCP Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9917R-A750
BOSSBS-9917R-A750EA 893 EUR InStock
BOSSBS-9917R-A750
Anti-PGCP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
PGCP is a 472 amino acid secreted protein that is primarily detected in blood plasma. PGCP is a carboxypeptidase that potentially is involved in the hydrolysis of circulating peptides. Due to its upregulation in hepatocellular carcinoma (HCC), it is suspected that PGCP may be a potential serological marker for HCC. PGCP is a member of the Peptidase M28 family of proteins, which also includes PSM (prostate-specific membrane antigen), metallopeptidases and aminopeptidases. The gene encoding PGCP maps to chromosome 8, which is made up of nearly 146 million bases and encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukaemias and lymphomas, and are typically associated with a poor prognosis. In humans, PGCP is found principally in blood plasma. It is a Carboxypeptidase that may play an important role in the hydrolysis of circulating peptides.

Type: Primary
Antigen: PGCP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-130/472
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge