Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C21orf91 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9983R-A750
BOSSBS-9983R-A750EA 893 EUR InStock
BOSSBS-9983R-A750
Anti-C21orf91 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
First is a 297 amino acid protein that contains a putative carboxy-terminal coiled-coil domain. With highest expression in embryonic dorsal retina and, during later embryonic stages, the anterior epithelial cells of the lens, it is suspected that EURL may play a role in cell determination and differentiation. The gene encoding EURL maps to the long arm of chromosome 21, which houses approximately 300 genes and comprises nearly 1.5% of the human genome. Chromosome 21-associated disorders include Alzheimer's Disease, amyotrophic lateral sclerosis and, most notably, Down Syndrome (also known as trisomy 21).

Type: Primary
Antigen: C21orf91
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/297
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge