Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-beta 2-MG Mouse Monoclonal Antibody [clone: B2E5] (ALEXA FLUOR® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BSM-0390M-A680
BOSSBSM-0390M-A680EA 1090 EUR InStock
BOSSBSM-0390M-A680
Anti-beta 2-MG Mouse Monoclonal Antibody [clone: B2E5] (ALEXA FLUOR® 680)
Antikörper
Beta 2 Microglobulin non-covalently associates with the 44 kDa alpha chain to form the HLA Class I antigen complex. Human beta 2 Microglobulin associated with HLA Class I antigens are expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes.

Type: Primary
Antigen: beta 2-MG
Clonality: Monoclonal
Clone: B2E5
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Mouse
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge