Cell disruptor with handheld converter and ¹/₈" microtip, Sonifier® SFX150
Lieferant: BRANSON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
101-063-1097R
BRAN101-063-1097REA 4769 EUR InStock
BRAN101-063-1097R
Cell disruptor with handheld converter and ¹/₈" microtip, Sonifier® SFX150
Homogenisatoren Ultraschall-Desintegratoren
The SFX150 Branson's handheld converter is designed with the user in mind with an ergonomic grip for comfort and control, and a recessed pushbutton and LED indicator for assured operation and to bring the industry's most advanced sample-processing capabilities to your laboratory.
  • Automatically adjust the ultrasonic pulse length
  • Prevent the sample from overheating (requires optional temperature probe)
  • Microtip limit 70%
  • Temperature control allows the user to specify a temperature range for the sample
Applications include sample preparation, cell disruption, nanoparticle disperson, mixing, homogenization, DNA, cell lysis, protein extration.
Geared toward low-volume samples ranging from 0,2 to 150 ml, the Sonifier SFX150 delivers up to 150 watts of power. And at 40 kHz, it's quiet enough to operate without an enclosure.
Advanced control modes includes time and energy mode, energy mode measured in joules, either continuously or in individual pulses. Process monitoring on a scrollable, digital screen provides updates of key variables including power level, energy usage, sample temperature, and experiment progress. For assured repeatability and precision on a greater scale, user can store up to 20 sample-processing programs. Program parameters include continuous or pulsed ultrasonics; time, energy, or temperature control modes; pulse on-time/energy, off-time and total on-time/energy; or amplitude as a percentage.
Zertifizierungen: CSA, UL, ISO9001, ISO14001.
Lieferumfang: Three year warranty. Includes 40 kHz handheld converter, ¹/₈" microtip, power cord, manual, wrench set. Other accessories also available.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge